missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methoxybenzoic acid, 98%

Product Code. 11906841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11906841 1 g 1g
11642179 5 g 5g
11926841 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11906841 Supplier Thermo Scientific Acros Supplier No. 437720010
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3177-80-8 | C8H9NO3 | 167.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3177-80-8
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.17
InChI Key SXOPCLUOUFQBJV-UHFFFAOYSA-N
Synonym 3-methoxyanthranilic acid, benzoic acid, 2-amino-3-methoxy, 3-methoxyanthranilate, m-anisic acid, 2-amino, 3-methoxy-2-aminobenzoic acid, 2-amino-3-methoxy-benzoic acid, 2-amino-3-methoxy benzoic acid, 2-amino-3-methyloxy benzoic acid, 2-amino-3-methoxybenzoicacid
PubChem CID 255720
ChEBI CHEBI:27440
IUPAC Name 2-amino-3-methoxybenzoic acid
SMILES COC1=CC=CC(=C1N)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 168°C to 170.7°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 167.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-3-methoxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-3-methoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.