missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 15413337
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15413337 10 g 10g
15403337 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15413337 Supplier Thermo Scientific Alfa Aesar Supplier No. H26001.09
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-18-8
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.135
MDL Number MFCD00024261
InChI Key JJPIVRWTAGQTPQ-UHFFFAOYSA-N
Synonym 2-amino-3-nitrobenzoic acid, 3-nitroanthranilic acid, benzoic acid, 2-amino-3-nitro, 2-amino-3-nitrobenzonic acid, 2-amino-3-nitro-benzoic acid, 2-amino-3-nitro benzoic acid, pubchem2233, amino-3-nitrobenzoic acid, 2-amino-nitro-benzoesaure, 2-carboxy-6-nitroaniline
PubChem CID 219633
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])N)C(=O)O

Specifications

Melting Point 210°C to 212°C
Quantity 10 g
Formula Weight 182.14
Percent Purity 97%
Chemical Name or Material 2-Amino-3-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.