missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Amino-4-carboxybenzeneboronic acid hydrochloride, 96%

Product Code. 15450176
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 15450176

Brand: Alfa Aesar™ H52888.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 850568-60-4
Melting Point >300°C
Molecular Formula C7H9BClNO4
MDL Number MFCD06656012
Quantity 250 mg
Synonym 3-amino-4-boronobenzoic acid hydrochloride, 2-amino-4-carboxylphenylboronic acid , hcl, 2-amino-4-carboxybenzeneboronic acid hydrochloride, 3-amino-4-dihydroxyboranyl benzoic acid hydrochloride, 3-amino-4-boronobenzoicacidhydrochloride, 2-amino-4-carboxylphenylboronic acid hcl, 3-amino-4-boronobenzoic acid, hcl, 3-amino-4-boronobenzoic acid, hydrochloride, 2-amino-4-carboxylphenylboronic acid hydrochloride, 2-amino-4-carboxy benzeneboronic acid hydrochloride
InChI Key ZRENJNMVSRNRES-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)C(=O)O)N)(O)O.Cl
IUPAC Name 3-amino-4-boronobenzoic acid;hydrochloride
Molecular Weight (g/mol) 217.412
PubChem CID 44118769
Formula Weight 217.42
Percent Purity 96%
Chemical Name or Material 2-Amino-4-carboxybenzeneboronic acid hydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.