missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorobenzoic acid, 98%

Product Code. 10607632 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10607632 25 g 25g
10488620 100 g 100g
10790565 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10607632 Supplier Thermo Scientific Acros Supplier No. 103320250
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89-77-0 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-77-0
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007778
InChI Key JYYLQSCZISREGY-UHFFFAOYSA-N
Synonym 4-chloroanthranilic acid, benzoic acid, 2-amino-4-chloro, 4-chloro-2-aminobenzoic acid, anthranilic acid, 4-chloro, 4-chloroanthranil acid, 4-chloro anthranilic acid, 2-carboxy-5-chloroaniline, unii-g55k85r12h, 2-amino-4-chloro-benzoic acid, 2,4-acba
PubChem CID 66646
IUPAC Name 2-amino-4-chlorobenzoic acid
SMILES NC1=CC(Cl)=CC=C1C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 231°C to 235°C
Color Beige to Brown
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula H2NC6H3(Cl)CO2H
Quantity 25 g
Beilstein 14, 365
Solubility Information Solubility in water: slightly soluble
Formula Weight 171.58
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-4-chlorobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-4-chlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.