missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-fluorobenzoic acid, 96%

Product Code. 10581544 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10581544 10 g 10g
10461101 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10581544 Supplier Thermo Scientific Acros Supplier No. 293020100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 446-32-2 | C7H6FNO2 | 155.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-32-2
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.13
MDL Number MFCD00075553
InChI Key LGPVTNAJFDUWLF-UHFFFAOYSA-N
Synonym 4-fluoroanthranilic acid, 2-amino-4-fluoro-benzoic acid, benzoic acid, 2-amino-4-fluoro, 2-amino-4-fluoro benzoic acid, 2-amino-4-fluorobenzoicacid, fa0, pubchem1323, amino-4-fluorobenzoic acid, acmc-209jz7, 2-amino-4fluorobenzoic acid
PubChem CID 2724967
IUPAC Name 2-amino-4-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)N)C(=O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 188.0°C to 196.0°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC6H3(F)COOH
Quantity 10 g
Formula Weight 155.13
Percent Purity 96%
Chemical Name or Material 2-Amino-4-fluorobenzoic acid
compliance-icons
Product Identifier
  • 2-Amino-4-fluorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.