missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-nitrophenol, 97%, Maybridge

Product Code. 10717305
Change view
Click to view available options
:
50g; 50g
Product Code.
10717305 50g; 50g
1 options
This item is not returnable. View return policy

Product Code. 10717305

Brand: Thermo Scientific Maybridge SB01488EE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 99-57-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H6N2O3
MDL Number MFCD00007695
Quantity 50 g
Synonym 2-hydroxy-5-nitroaniline, phenol, 2-amino-4-nitro, 4-nitro-2-aminophenol, p-nitro-o-aminophenol, 4-nitro-2-aminofenol, p-nitroaminofenol, 3-amino-4-hydroxynitrobenzene, 1-nitro-3-amino-4-hydroxybenzene, rodol 42, 2-amino-4-nitrofenol
InChI Key VLZVIIYRNMWPSN-UHFFFAOYSA-N
SMILES NC1=CC(=CC=C1O)[N+]([O-])=O
IUPAC Name 2-amino-4-nitrophenol
Molecular Weight (g/mol) 154.13
PubChem CID 3613389
ChEBI CHEBI:82383
Formula Weight 154.12
Percent Purity 97%
Chemical Name or Material 2-Amino-4-nitrophenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.