missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-nitrophenol, 98%, Thermo Scientific Chemicals

Product Code. 11471718
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471718 500 g 500g
11451718 25 g 25g
11461718 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471718 Supplier Thermo Scientific Alfa Aesar Supplier No. A13964.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-57-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00007695
InChI Key VLZVIIYRNMWPSN-UHFFFAOYSA-N
Synonym 2-hydroxy-5-nitroaniline, phenol, 2-amino-4-nitro, 4-nitro-2-aminophenol, p-nitro-o-aminophenol, 4-nitro-2-aminofenol, p-nitroaminofenol, 3-amino-4-hydroxynitrobenzene, 1-nitro-3-amino-4-hydroxybenzene, rodol 42, 2-amino-4-nitrofenol
PubChem CID 3613389
ChEBI CHEBI:82383
IUPAC Name 2-amino-4-nitrophenol
SMILES NC1=CC(=CC=C1O)[N+]([O-])=O

Specifications

Melting Point 141°C to 145°C
Quantity 500 g
Beilstein 776533
Formula Weight 154.13
Percent Purity 98%
Chemical Name or Material 2-Amino-4-nitrophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.