missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-phenylphenol, 98%, Thermo Scientific™

Product Code. 11995311 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11995311 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11995311 Supplier Thermo Scientific Acros Supplier No. 439930010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1134-36-7
CAS Min % 97.5
CAS Max % 100.0
Melting Point 208.5°C to 209°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C12H11NO
Quantity 1 g
Synonym 3-amino-4-hydroxybiphenyl, 3-aminobiphenyl-4-ol, 3-amino-1,1'-biphenyl-4-ol, 2-hydroxy-5-phenylaniline, 4-biphenylol, 3-amino, 1,1'-biphenyl-4-ol, 3-amino, phenol, 2-amino-4-phenyl, 2-amino-4-phenyl-phenol, 2-azanyl-4-phenyl-phenol
InChI Key IGIDZGNPFWGICD-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=CC(=C(C=C2)O)N
IUPAC Name 2-amino-4-phenylphenol
Molecular Weight (g/mol) 185.22
PubChem CID 14562
Formula Weight 185.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-4-phenylphenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-4-phenylphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.