missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(trifluoromethyl)thiophenol hydrochloride, 97%

Product Code. 11362207
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11362207

Brand: Thermo Scientific Alfa Aesar L09683.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4274-38-8 | C7H7ClF3NS | 229.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4274-38-8
Molecular Formula C7H7ClF3NS
Molecular Weight (g/mol) 229.65
MDL Number MFCD00042600
InChI Key FIAGYDIJZOWVAB-UHFFFAOYSA-N
Synonym 2-amino-4-trifluoromethyl benzenethiol hydrochloride, 3-amino-4-mercaptobenzotrifluoride hydrochloride, 2-amino-4-trifluoromethylthiophenol hydrochloride, 2-amino-4-trifluoromethyl benzene-1-thiol hydrochloride, 2-amino-4-trifluoromethyl thiophenol hydrochloride, 2-amino-4-trifluoromethyl benzene-1-thiol, chloride, pubchem2827, c7h6f3ns.hcl, dsstox_cid_31681, dsstox_rid_97532
PubChem CID 2734678
IUPAC Name 2-amino-4-(trifluoromethyl)benzenethiol;hydrochloride
SMILES [H+].[Cl-].NC1=CC(=CC=C1S)C(F)(F)F

Specifications

Melting Point 186°C to 190°C
Density 2.46
Quantity 1 g
UN Number UN1759
Beilstein 4273774
Formula Weight 229.65
Percent Purity 97%
Chemical Name or Material 2-Amino-4-(trifluoromethyl)thiophenol hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.