missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-2'-fluorobenzophenone, 95%, Thermo Scientific Chemicals

Product Code. 15418478
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418478 5 g 5g
15408478 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418478 Supplier Thermo Scientific Alfa Aesar Supplier No. H61882.06
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1479-58-9 | Molecular Formula: C13H9BrFNO | Molecular Weight: 294.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1479-58-9
Molecular Formula C13H9BrFNO
Molecular Weight (g/mol) 294.12
MDL Number MFCD00038380
InChI Key XCOKDXNGCQXFCV-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-2'-fluorobenzophenone, 2-amino-2'-fluoro-5-bromobenzophenone, 2-amino-5-bromophenyl 2-fluorophenyl methanone, methanone, 2-amino-5-bromophenyl 2-fluorophenyl, 4-bromo-2-2-fluorobenzoyl aniline, 2-amino-5-bromophenyl 2-fluorophenyl ketone, 2-amino-5-bromo-2-fluorobenzophenone, 2-amino-5-bromo-phenyl-2-fluoro-phenyl-methanone, 2-amino-2-fluoro-5-bromobenzophenone, 2-amino-5-bromo-2'-fluoro benzophenone
PubChem CID 73865
IUPAC Name (2-amino-5-bromophenyl)-(2-fluorophenyl)methanone
SMILES NC1=CC=C(Br)C=C1C(=O)C1=CC=CC=C1F

Specifications

Melting Point 100°C to 104°C
Quantity 5 g
Formula Weight 294.12
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-5-bromo-2'-fluorobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.