missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromobenzoic acid, 98%

Product Code. 11453894
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453894 5 g 5g
11463894 25 g 25g
11473894 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453894 Supplier Thermo Scientific Alfa Aesar Supplier No. A11915.06
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5794-88-7 | C7H6BrNO2 | 216.034 g/mol

It is used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. Also used in the determination of cobalt, copper, nickel, and zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. Also used in the determination of cobalt, copper, nickel, and zinc.

Solubility
Soluble in methanol and dimethyl sulfoxide.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5794-88-7
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00007823
InChI Key CUKXRHLWPSBCTI-UHFFFAOYSA-N
Synonym 5-bromoanthranilic acid, benzoic acid, 2-amino-5-bromo, anthranilic acid, 5-bromo, 2-amino-5-bromo-benzoic acid, 5-bromo-2-aminobenzoic acid, 2-amino-5-bromobenzoicacid, 2-amino-5-bromo benzoic acid, 5-bromo anthranilic acid, buttpark 89\07-49, 5-bromo-2-amino-benzoic acid
PubChem CID 79858
IUPAC Name 2-amino-5-bromobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)N

Specifications

Melting Point ∼215°C (decomposition)
Quantity 5 g
Beilstein 639028
Sensitivity Light sensitive
Merck Index 14,1405
Solubility Information Soluble in methanol and dimethyl sulfoxide.
Formula Weight 216.03
Percent Purity 98%
Chemical Name or Material 2-Amino-5-bromobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.