missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzoic acid, 98%

Product Code. 11446887
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446887 100 g 100g
11426887 5 g 5g
11436887 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446887 Supplier Thermo Scientific Alfa Aesar Supplier No. A14329.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 635-21-2 | C7H6ClNO2 | 171.58 g/mol

2-Amino-5-chlorobenzoic acid is used in the preparation of disease-modifying antirheumatic drugs (DMARDs). Also used to produce 6-chloro-3H-quinazolin-4-one at temperature of 180°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-chlorobenzoic acid is used in the preparation of disease-modifying antirheumatic drugs (DMARDs). Also used to produce 6-chloro-3H-quinazolin-4-one at temperature of 180°C.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-21-2
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007838
InChI Key IFXKXCLVKQVVDI-UHFFFAOYSA-N
Synonym 5-chloroanthranilic acid, 5-chloro-2-aminobenzoic acid, benzoic acid, 2-amino-5-chloro, anthranilic acid, 5-chloro, 2-amino-5-chloro-benzoic acid, unii-z2y66jmi31, 2-carboxy-4-chloroaniline, chembl36767, 5-amino-3-carboxy-1-chlorobenzene, 2-amino-5-chlorobenzoicacid
PubChem CID 12476
IUPAC Name 2-amino-5-chlorobenzoic acid
SMILES C1=CC(=C(C=C1Cl)C(=O)O)N

Specifications

Melting Point ∼208°C (decomposition)
Quantity 100 g
Beilstein 639027
Solubility Information Soluble in water.
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 2-Amino-5-chlorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.