missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzophenone, 98+%

Product Code. 11417707
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417707 100 g 100g
11427707 500 g 500g
11437707 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11417707 Supplier Thermo Scientific Alfa Aesar Supplier No. A15720.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 719-59-5 | C13H10ClNO | 231.68 g/mol

It is used in the heterocyclic syntheses based on the reactions of dimethyl acetylenedicarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the heterocyclic syntheses based on the reactions of dimethyl acetylenedicarboxylate.

Solubility
Solubility in methanol gives very faint turbidity. Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 719-59-5
Molecular Formula C13H10ClNO
Molecular Weight (g/mol) 231.68
MDL Number MFCD00007839
InChI Key ZUWXHHBROGLWNH-UHFFFAOYSA-N
Synonym 2-amino-5-chlorobenzophenone, 2-amino-5-chlorophenyl phenyl methanone, 2-benzoyl-4-chloroaniline, methanone, 2-amino-5-chlorophenyl phenyl, 2-amino-5-chlorbenzophenone, 5-chloro-2-aminobenzophenone, 2-amino-5-chlorobenzylphenone, benzophenone, 2-amino-5-chloro, unii-fr80014zbt, 2-amino-5-chlorophenyl phenylmethanone
PubChem CID 12870
IUPAC Name (2-amino-5-chlorophenyl)-phenylmethanone
SMILES NC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1

Specifications

Melting Point 96°C to 99°C
Density 1.33
Flash Point 211°C (412°F)
Quantity 100 g
Beilstein 475640
Formula Weight 231.68
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-chlorobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.