missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-chlorobenzophenone, 98+%
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
Description
It is used in the heterocyclic syntheses based on the reactions of dimethyl acetylenedicarboxylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the heterocyclic syntheses based on the reactions of dimethyl acetylenedicarboxylate.
Solubility
Solubility in methanol gives very faint turbidity. Soluble in DMSO.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used in the heterocyclic syntheses based on the reactions of dimethyl acetylenedicarboxylate.
Solubility
Solubility in methanol gives very faint turbidity. Soluble in DMSO.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 96°C to 99°C |
| Density | 1.33 |
| Flash Point | 211°C (412°F) |
| Quantity | 500 g |
| Beilstein | 475640 |
| Formula Weight | 231.68 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 2-Amino-5-chlorobenzophenone |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction