missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzophenone, TRC

Product Code. 30132488
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132488 50 g 50g
17953485 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132488 Supplier Toronto Research Chemicals Supplier No. TRCA60349050G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 719-59-5
Molecular Formula C13 H10 Cl N O
Molecular Weight (g/mol) 231.68
Synonym (2-Amino-5-chlorophenyl)phenylmethanone, Prazepam Imp. C (EP), Dipotassium Clorazepate Imp. A (EP), Alprazolam Imp. E (EP), Oxazepam Imp. D (EP), 2-Amino-5-chlorobenzophenone, Chlordiazepoxide Hydrochloride Imp. C (EP), Aminochlorobenzophenone
IUPAC Name (2-amino-5-chlorophenyl)-phenylmethanone
SMILES Nc1ccc(Cl)cc1C(=O)c2ccccc2

Specifications

Quantity 50 g
InChI Formula InChI=1S/C13H10ClNO/c14-10-6-7-12(15)11(8-10)13(16)9-4-2-1-3-5-9/h1-8H,15H2
Formula Weight 231.0451
Percent Purity >95%
Chemical Name or Material 2-Amino-5-chlorobenzophenone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.