missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorophenol, 97%

Product Code. 15422419
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15422419 25 g 25g
15432419 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422419 Supplier Thermo Scientific Alfa Aesar Supplier No. H64978.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28443-50-7 | C6H6ClNO | 143.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28443-50-7
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD02093863
InChI Key FZCQMIRJCGWWCL-UHFFFAOYSA-N
Synonym 4-chloro-2-hydroxyaniline, phenol, 2-amino-5-chloro, 2-amino-5-chloro-phenol, 5-chloro-2-aminophenol, pubchem1478, 5-chloro-o-aminophenol, 5-chloro-2-amino phenol, 2-hydroxy-4-chloroaniline, acmc-209h2j, 2-azanyl-5-chloranyl-phenol
PubChem CID 91591
ChEBI CHEBI:75051
IUPAC Name 2-amino-5-chlorophenol
SMILES NC1=CC=C(Cl)C=C1O

Specifications

Melting Point 145°C to 153°C
Color Brown
Quantity 25 g
UN Number UN2811
Beilstein 386421
Formula Weight 143.57
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 2-Amino-5-chlorophenol
compliance-icons
Product Identifier
  • 2-Amino-5-chlorophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Amino-5-chlorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.