missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluorobenzothiazole, 98%

Product Code. 11416700
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416700 1 g 1g
15457907 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416700 Supplier Thermo Scientific Alfa Aesar Supplier No. H51022.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20358-07-0 | C7H5FN2S | 168.189 g/mol

2-Amino-5-fluorobenzothiazole is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-fluorobenzothiazole is used as organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20358-07-0
Molecular Formula C7H5FN2S
Molecular Weight (g/mol) 168.189
MDL Number MFCD05664560
InChI Key YHBIGBYIUMCLJS-UHFFFAOYSA-N
PubChem CID 821203
IUPAC Name 5-fluoro-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1F)N=C(S2)N

Specifications

Quantity 1 g
Synonym 2-amino-5-fluorobenzothiazole, 5-fluorobenzo d thiazol-2-amine, 2-benzothiazolamine, 5-fluoro, 5-fluoro-2-benzothiazolamine, 2-benzothiazolamine,5-fluoro, 5-fluorobenzothiazol-2-ylamine, pubchem21850, acmc-20a67y, 2-amino-5-fluorobenzothiozole, 5-fluoro-2-aminobenzothiazole
Solubility Information Soluble in methanol.
Formula Weight 168.19
Percent Purity 98%
Chemical Name or Material 2-Amino-5-fluorobenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.