missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-nitrobenzotrifluoride, 98%, Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsSpecifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 90°C to 93°C |
| Color | Yellow to Brown |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Molecular Formula | C7H5F3N2O2 |
| Linear Formula | H2NC6H3(NO2)CF3 |
| Quantity | 5 g |
| Show More |
Safety and Handling
Product Identifier
- 2-Amino-5-nitrobenzotrifluoride
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?