missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-phenyl-3-furonitrile, 95%, Thermo Scientific™

Product Code. 10763613
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10763613 1 g Amber Glass Bottle 1g
10773613 10 g Amber Glass Bottle 10g
10183444 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10763613 Supplier Thermo Scientific Maybridge Supplier No. AW01254DA
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 14742-32-6
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.20
MDL Number MFCD00463472
InChI Key UWTDTJWGYWUILP-UHFFFAOYSA-N
Synonym 2-amino-5-phenyl-3-furonitrile, 2-amino-3-cyano-5-phenylfuran, 2-amino-5-phenyl-3-furancarbonitrile, 3-furancarbonitrile, 2-amino-5-phenyl, maybridge1_000452, 2-amino-5-phenyl-furan-3-carbonitrile, 3-furancarbonitrile,2-amino-5-phenyl, 2-azanyl-5-phenyl-furan-3-carbonitrile
PubChem CID 693961
IUPAC Name 2-amino-5-phenylfuran-3-carbonitrile
SMILES NC1=C(C=C(O1)C1=CC=CC=C1)C#N

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 184.2
Percent Purity 95%
Chemical Name or Material 2-Amino-5-phenyl-3-furonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.