missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-(trifluoromethyl)benzoic acid, 95%

Product Code. 15491209
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491209 1 g 1g
15401219 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15491209 Supplier Thermo Scientific Alfa Aesar Supplier No. H64141.03
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 83265-53-6 | C8H6F3NO2 | 205.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83265-53-6
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.136
MDL Number MFCD01569537
InChI Key GLCQUPLYYXSPQB-UHFFFAOYSA-N
Synonym 2-amino-5-trifluoromethyl benzoic acid, 2-amino-5-trifluoromethyl-benzoic acid, 5-trifluoromethylanthranilic acid, 5-trifluoromethyl anthranilic acid, benzoic acid, 2-amino-5-trifluoromethyl, 2-amino-5-trifluoromethyl-benzoicacid, pubchem12904, 4-amino-3-carboxybenzotrifluoride, 2-carboxy-4-trifluoromethyl aniline, 2-amino-5-trifluoromethyl benzoicacid
PubChem CID 10465535
IUPAC Name 2-amino-5-(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C=C1C(F)(F)F)C(=O)O)N

Specifications

Melting Point 141°C to 146°C (decomposition)
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 205.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-5-(trifluoromethyl)benzoic acid
compliance-icons
Product Identifier
  • 2-Amino-5-(trifluoromethyl)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Amino-5-(trifluoromethyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.