missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-bromobenzoic acid, 97+%

Product Code. 15434787
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15434787 5 g 5g
15424787 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15434787 Supplier Thermo Scientific Alfa Aesar Supplier No. H32106.06
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20776-48-1 | C7H6BrNO2 | 216.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20776-48-1
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD03618455
InChI Key BNQPROAXWQCNKO-UHFFFAOYSA-N
Synonym 6-bromoanthranilic acid, 3-bromo-2-carboxyaniline, benzoic acid, 2-amino-6-bromo, buttpark 49\07-50, 2-amino-6-bromo-benzoic acid, ksc201s8r
PubChem CID 10560649
IUPAC Name 2-amino-6-bromobenzoic acid
SMILES NC1=C(C(O)=O)C(Br)=CC=C1

Specifications

Quantity 5 g
Sensitivity Light sensitive
Formula Weight 216.03
Percent Purity 98%
Chemical Name or Material 2-Amino-6-bromobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.