missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methoxybenzoic acid, 97%

Product Code. 15427057
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15427057 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15427057 Supplier Thermo Scientific Alfa Aesar Supplier No. H34128.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 53600-33-2 | C8H9NO3 | 167.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 53600-33-2
Melting Point ∽85°C (decomposition)
Molecular Formula C8H9NO3
MDL Number MFCD01941328
Quantity 1 g
Synonym 6-methoxyanthranilic acid, 6-amino-o-anisic acid, 2-amino-6-methoxy-benzoic acid, 2-amino-6-methoxybenzoicacid, benzoic acid, 2-amino-6-methoxy, 6-methoxy anthranilic acid, pubchem4706, 3-amino-2-carboxyanisole, 6-methoxy-anthranilic acid, 2-carboxy-3-methoxyaniline
InChI Key DYZDIWNRWSNVPT-UHFFFAOYSA-N
SMILES COC1=CC=CC(=C1C(=O)O)N
IUPAC Name 2-amino-6-methoxybenzoic acid
Molecular Weight (g/mol) 167.164
PubChem CID 2735357
Formula Weight 167.16
Percent Purity 97%
Chemical Name or Material 2-Amino-6-methoxybenzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.