missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methylbenzoic acid, 98%

Product Code. 11472198
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472198 100 g 100g
11452198 5 g 5g
11462198 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472198 Supplier Thermo Scientific Alfa Aesar Supplier No. A15187.22
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4389-50-8 | C8H8NO2 | 150.16 g/mol

2-Amino-6-methylbenzoic acid reacts with cyanogen bromide to prepare 2-amino-5-methyl-benzo[d][1,3]oxazin-4-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-methylbenzoic acid reacts with cyanogen bromide to prepare 2-amino-5-methyl-benzo[d][1,3]oxazin-4-one.

Solubility
Soluble in methanol.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4389-50-8
Molecular Formula C8H8NO2
Molecular Weight (g/mol) 150.16
MDL Number MFCD00007809
InChI Key XHYVBIXKORFHFM-UHFFFAOYSA-M
Synonym 6-methylanthranilic acid, 6-amino-o-toluic acid, benzoic acid, 2-amino-6-methyl, 2-amino-6-methyl-benzoic acid, 6-methylanthranilate, pubchem4957, acmc-209jvt, 6-methyl anthranilic acid, 3-amino-2-carboxytoluene, 2-carboxy-3-methylaniline
PubChem CID 151210
IUPAC Name 2-amino-6-methylbenzoic acid
SMILES CC1=CC=CC(N)=C1C([O-])=O

Specifications

Melting Point ∼127°C (decomposition)
Quantity 100 g
Beilstein 2803391
Solubility Information Soluble in methanol.
Formula Weight 151.16
Percent Purity 98%
Chemical Name or Material 2-Amino-6-methylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.