missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methylbenzoic acid, 98%

Product Code. p-7022418
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11452198

Brand: Thermo Scientific Alfa Aesar A15187.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4389-50-8 | C8H8NO2 | 150.16 g/mol

2-Amino-6-methylbenzoic acid reacts with cyanogen bromide to prepare 2-amino-5-methyl-benzo[d][1,3]oxazin-4-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-methylbenzoic acid reacts with cyanogen bromide to prepare 2-amino-5-methyl-benzo[d][1,3]oxazin-4-one.

Solubility
Soluble in methanol.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4389-50-8
Molecular Formula C8H8NO2
Molecular Weight (g/mol) 150.16
MDL Number MFCD00007809
InChI Key XHYVBIXKORFHFM-UHFFFAOYSA-M
Synonym 6-methylanthranilic acid, 6-amino-o-toluic acid, benzoic acid, 2-amino-6-methyl, 2-amino-6-methyl-benzoic acid, 6-methylanthranilate, pubchem4957, acmc-209jvt, 6-methyl anthranilic acid, 3-amino-2-carboxytoluene, 2-carboxy-3-methylaniline
PubChem CID 151210
IUPAC Name 2-amino-6-methylbenzoic acid
SMILES CC1=CC=CC(N)=C1C([O-])=O

Specifications

Melting Point ∼127°C (decomposition)
Quantity 5 g
Beilstein 2803391
Solubility Information Soluble in methanol.
Formula Weight 151.16
Percent Purity 98%
Chemical Name or Material 2-Amino-6-methylbenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.