missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals

Product Code. 15492109
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492109 5 g 5g
15402119 250 mg 250mg
15482109 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492109 Supplier Thermo Scientific Alfa Aesar Supplier No. H64771.06
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108714-73-4
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.292
MDL Number MFCD09953790
InChI Key GUTJITRKAMCHSD-UHFFFAOYSA-N
Synonym 2-amino-9,9-dimethylfluorene, 9,9-dimethyl-9h-fluoren-2-amine, 2-amino-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-9h-fluoren-2-ylamine, 9,9-dimethyl9h-fluorene-2-amine, pubchem23234, ksc495c6d, 9,9-dimethyl-2-aminofluorene, 2-amino-9,9-dimethyl fluorene, 9,9-dimethyl-fluoren-2-amine
PubChem CID 22617093
IUPAC Name 9,9-dimethylfluoren-2-amine
SMILES CC1(C2=CC=CC=C2C3=C1C=C(C=C3)N)C

Specifications

Color White
Quantity 5 g
Beilstein 15506865
Formula Weight 209.29
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-9,9-dimethylfluorene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.