missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-9-fluorenone, 98%

Product Code. 15471168
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15471168 5 g 5g
15461168 1 g 1g
15481168 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15471168 Supplier Thermo Scientific Alfa Aesar Supplier No. H55048.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3096-57-9 | C13H9NO | 195.22 g/mol

2-Amino-9-fluorenone is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-9-fluorenone is used as an active pharmaceutical intermediate.

Solubility
Soluble in alcohol and ether. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3096-57-9
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.22
MDL Number MFCD00001157
InChI Key SJODITPGMMSNRF-UHFFFAOYSA-N
Synonym 2-amino-9-fluorenone, 2-amino-9h-fluoren-9-one, 2-aminofluorenone, 9h-fluoren-9-one, 2-amino, 9h-fluoren-9-one,2-amino, fluoren-9-one, 2-amino, 9h-fluoren-9-one, 2-amino-9ci, 2-amino-fluorenon, 2-aminofluorenone-9, 2-aminofluorene-9-one
PubChem CID 18357
IUPAC Name 2-aminofluoren-9-one
SMILES NC1=CC=C2C3=CC=CC=C3C(=O)C2=C1

Specifications

Melting Point 157°C to 160°C
Quantity 5 g
Solubility Information Soluble in alcohol and ether. Insoluble in water.
Formula Weight 195.22
Percent Purity 98%
Chemical Name or Material 2-Amino-9-fluorenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.