missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-N-(2-chloro-6-methylphenyl)-5-thiazolecarboxamide, TRC

Product Code. 30134004
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134004 10 g 10g
30012146 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134004 Supplier Toronto Research Chemicals Supplier No. TRCA60391010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302964-24-5
Molecular Formula C11 H10 Cl N3 O S
Molecular Weight (g/mol) 267.73
Synonym 5-Thiazolecarboxamide, 2-amino-N-(2-chloro-6-methylphenyl)-, 2-Amino-N-(2-chloro-6-methylphenyl)-5-thiazolecarboxamide, 2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide
IUPAC Name 2-amino-N-(2-chloro-6-methylphenyl)-1,3-thiazole-5-carboxamide
SMILES Cc1cccc(Cl)c1NC(=O)c2cnc(N)s2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H10ClN3OS/c1-6-3-2-4-7(12)9(6)15-10(16)8-5-14-11(13)17-8/h2-5H,1H3,(H2,13,14)(H,15,16)
Formula Weight 267.0233
Chemical Name or Material 2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.