missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-p-cresol, 97%

Product Code. 10861861 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10861861 50 g Glass bottle 50g
10255830 250 g Glass bottle 250g
10371620 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10861861 Supplier Thermo Scientific Acros Supplier No. 163210500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 95-84-1 | C7H9NO | 123.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-84-1
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00007699
InChI Key ZMXYNJXDULEQCK-UHFFFAOYSA-N
Synonym 2-amino-p-cresol, 2-hydroxy-5-methylaniline, phenol, 2-amino-4-methyl, 3-amino-4-hydroxytoluene, 6-hydroxy-m-toluidine, 4-methyl-2-aminophenol, 5-methyl-2-hydroxyaniline, 2-amino-4-cresol, p-cresol, 2-amino, 2-amino-4-methyl-phenol
PubChem CID 7264
IUPAC Name 2-amino-4-methylphenol
SMILES CC1=CC=C(O)C(N)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 134.0°C to 139.0°C
Color Beige to Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H3(CH3)OH
Quantity 50 g
Beilstein 13,601
Solubility Information Solubility in water: slightly soluble
Formula Weight 123.16
Percent Purity 97%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 2-Amino-p-cresol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-p-cresol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.