missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzamide, 98+%

Product Code. 11477806
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477806 500 g 500g
11467806 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477806 Supplier Thermo Scientific Alfa Aesar Supplier No. A14756.36
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 88-68-6 | C7H8N2O | 136.154 g/mol

2-Aminobenzamide is used as an acetyldehyde scavenger in PET bottles. It is also used as an intermediate for ag chemicals, dyes, pharmaceuticals, blowing agents, and fine organic chemicals. In lubricating oils for supersonic jet engines, inhibits copper and magnesium corrosion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzamide is used as an acetyldehyde scavenger in PET bottles. It is also used as an intermediate for ag chemicals, dyes, pharmaceuticals, blowing agents, and fine organic chemicals. In lubricating oils for supersonic jet engines, inhibits copper and magnesium corrosion.

Solubility
Soluble in hot water and alcohol. Slightly soluble in ether and benzene. Very soluble in ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-68-6
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00007981
InChI Key PXBFMLJZNCDSMP-UHFFFAOYSA-N
Synonym anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van
PubChem CID 6942
IUPAC Name 2-aminobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)N

Specifications

Melting Point 110°C to 114°C
Boiling Point 300°C
Flash Point 198°C (388°F)
Quantity 500 g
Beilstein 508509
Solubility Information Soluble in hot water and alcohol. Slightly soluble in ether and benzene. Very soluble in ethyl acetate.
Formula Weight 136.15
Percent Purity ≥98%
Chemical Name or Material 2-Aminobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.