missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Aminobenzanilide, 95%

Product Code. 11357776
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357776 25 g 25g
11347776 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357776 Supplier Alfa Aesar™ Supplier No. L11451.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4424-17-3
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00034792,MFCD00017096
InChI Key FDPVTENMNDHFNK-UHFFFAOYSA-N
Synonym 2-aminobenzanilide, benzamide, 2-amino-n-phenyl, anthranilanilide, n-2-aminobenzoyl aniline, n-phenylanthranilamide, benzanilide, 2-amino, n-phenyl-2-aminobenzamide, 2-amino-n-phenyl-benzamide, 2-aminophenyl-n-benzamide, o-aminobenzanilide
PubChem CID 78142
IUPAC Name 2-amino-N-phenylbenzamide
SMILES NC1=CC=CC=C1C(=O)NC1=CC=CC=C1

Specifications

Melting Point 130°C to 134°C
Quantity 25 g
Beilstein 782786
Formula Weight 212.25
Percent Purity 95%
Chemical Name or Material 2-Aminobenzanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.