missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Aminobenzanilide, 95%

Product Code. p-7045637
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11347776

Brand: Alfa Aesar™ L11451.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4424-17-3
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00034792,MFCD00017096
InChI Key FDPVTENMNDHFNK-UHFFFAOYSA-N
Synonym 2-aminobenzanilide, benzamide, 2-amino-n-phenyl, anthranilanilide, n-2-aminobenzoyl aniline, n-phenylanthranilamide, benzanilide, 2-amino, n-phenyl-2-aminobenzamide, 2-amino-n-phenyl-benzamide, 2-aminophenyl-n-benzamide, o-aminobenzanilide
PubChem CID 78142
IUPAC Name 2-amino-N-phenylbenzamide
SMILES NC1=CC=CC=C1C(=O)NC1=CC=CC=C1

Specifications

Melting Point 130°C to 134°C
Quantity 5 g
Beilstein 782786
Formula Weight 212.25
Percent Purity 95%
Chemical Name or Material 2-Aminobenzanilide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.