missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzenesulfonamide, MedChemExpress

Product Code. 16408856
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16408856 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408856 Supplier Medchem Express Supplier No. HYB2147
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2-Aminobenzenesulfonamide is a carbonic anhydrase IX inhibitor.

2-Aminobenzenesulfonamide is a carbonic anhydrase IX inhibitor.

Specifications

Chemical Name or Material 2-Aminobenzenesulfonamide
CAS 3306-62-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C6H8N2O2S
Quantity 100 mg
Synonym Orthanilamide
Solubility Information DMSO : 160 mg/mL (929.15 mM; Need ultrasonic and warming)
SMILES O=S(C1=CC=CC=C1N)(N)=O
Molecular Weight (g/mol) 172.2
Formula Weight 172.2
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.