missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzophenone, 98%

Product Code. 10637052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10637052 25 g Glass bottle 25g
10647052 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10637052 Supplier Thermo Scientific Acros Supplier No. 103160250
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2835-77-0 | C13H11NO | 197.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-77-0
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00007713
InChI Key MAOBFOXLCJIFLV-UHFFFAOYSA-N
Synonym 2-aminobenzophenone, 2-benzoylaniline, o-benzoylaniline, o-aminobenzophenone, 2-aminophenyl phenyl methanone, methanone, 2-aminophenyl phenyl, benzophenone, 2-amino, 2-amino-phenyl-phenyl-methanone, 2-aminophenyl phenyl ketone, methanone, aminophenyl phenyl
PubChem CID 76080
IUPAC Name (2-aminophenyl)-phenylmethanone
SMILES NC1=CC=CC=C1C(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 105.0°C to 108.0°C
Color Golden-Yellow
Flash Point 240°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC6H4COC6H5
Quantity 25 g
Beilstein 14, 76
Solubility Information Solubility in water: practically insoluble. Other solubilities: 40g/L methanol (20°C),soluble in chloroform
Formula Weight 197.24
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Aminobenzophenone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminobenzophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.