missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzophenone, 98%

Product Code. 10647052 Shop All Thermo Scientific Chemicals Products
Cambiar vista
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Tamaño de la unidad:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging Tamaño de la unidad
10647052 100 g Glass bottle 100g
10637052 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10647052 Proveedor Thermo Scientific Acros N.º de proveedor 103161000
ChemsChrom Mega Deals
Use promo code 27993
Ver todas las promociones
 Restricted Item
Estimated Shipment: 27-11-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 2835-77-0 | C13H11NO | 197.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 2835-77-0
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00007713
InChI Key MAOBFOXLCJIFLV-UHFFFAOYSA-N
Synonym 2-aminobenzophenone, 2-benzoylaniline, o-benzoylaniline, o-aminobenzophenone, 2-aminophenyl phenyl methanone, methanone, 2-aminophenyl phenyl, benzophenone, 2-amino, 2-amino-phenyl-phenyl-methanone, 2-aminophenyl phenyl ketone, methanone, aminophenyl phenyl
PubChem CID 76080
IUPAC Name (2-aminophenyl)-phenylmethanone
SMILES NC1=CC=CC=C1C(=O)C1=CC=CC=C1

Especificaciones

CAS Min % 97.5
CAS Max % 100.0
Melting Point 105.0°C to 108.0°C
Color Golden-Yellow
Flash Point 240°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC6H4COC6H5
Quantity 100 g
Beilstein 14, 76
Solubility Information Solubility in water: practically insoluble. Other solubilities: 40g/L methanol (20°C),soluble in chloroform
Formula Weight 197.24
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Aminobenzophenone, 98%
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 2-Aminobenzophenone
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.