missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzophenone, 98%

Product Code. 11494284
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494284 10 g 10g
11404294 50 g 50g
11414294 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494284 Supplier Thermo Scientific Alfa Aesar Supplier No. A12580.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2835-77-0 | C13H11NO | 197.24 g/mol

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

Solubility
Insoluble in water. Soluble in DMSO, Methanol.

Notes
Store at room temperature. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2835-77-0
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00007713
InChI Key MAOBFOXLCJIFLV-UHFFFAOYSA-N
Synonym 2-aminobenzophenone, 2-benzoylaniline, o-benzoylaniline, o-aminobenzophenone, 2-aminophenyl phenyl methanone, methanone, 2-aminophenyl phenyl, benzophenone, 2-amino, 2-amino-phenyl-phenyl-methanone, 2-aminophenyl phenyl ketone, methanone, aminophenyl phenyl
PubChem CID 76080
IUPAC Name (2-aminophenyl)-phenylmethanone
SMILES NC1=CC=CC=C1C(=O)C1=CC=CC=C1

Specifikationer

Melting Point 105°C to 108°C
Boiling Point 280°C
Flash Point 240°C (464°F)
Quantity 10 g
Beilstein 743425
Solubility Information Insoluble in water. Soluble in DMSO,Methanol.
Formula Weight 197.24
Percent Purity 98%
Chemical Name or Material 2-Aminobenzophenone

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.