missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobiphenyl, 97%

Product Code. 11387575
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387575 1 g 1g
11397575 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387575 Supplier Thermo Scientific Alfa Aesar Supplier No. L05728.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 90-41-5 | C12H11N | 169.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-41-5
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00007701
InChI Key TWBPWBPGNQWFSJ-UHFFFAOYSA-N
Synonym 2-aminobiphenyl, 1,1'-biphenyl-2-amine, 2-biphenylamine, 2-aminodiphenyl, o-aminobiphenyl, o-biphenylamine, biphenyl-2-ylamine, o-aminodiphenyl, o-phenylaniline, biphenyl-2-amine
PubChem CID 7015
IUPAC Name 2-phenylaniline
SMILES NC1=CC=CC=C1C1=CC=CC=C1

Specifications

Melting Point 47°C to 52°C
Boiling Point 299°C
Flash Point 153°C (307°F)
Quantity 1 g
Beilstein 471874
Formula Weight 169.23
Percent Purity 97%
Chemical Name or Material 2-Aminobiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.