missing translation for 'onlineSavingsMsg'
Learn More

2-Aminonaphthalene-1-sulfonic acid, 97%

Product Code. 11432938
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432938 500 g 500g
11442938 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432938 Supplier Thermo Scientific Alfa Aesar Supplier No. A18644.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 81-16-3 | C10H9NO3S | 223.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-16-3
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.246
MDL Number MFCD00003988
InChI Key GWIAAIUASRVOIA-UHFFFAOYSA-N
Synonym tobias acid, 2-amino-1-naphthalenesulfonic acid, 2-naphthylamine-1-sulfonic acid, 1-naphthalenesulfonic acid, 2-amino, kyselina tobiasova, unii-f9qkw1fce0, kyselina 2-naftylamin-1-sulfonova, kyselina tobiasova czech, ccris 9048, 2-aminonaphthalene-1-sulphonic acid
PubChem CID 6670
IUPAC Name 2-aminonaphthalene-1-sulfonic acid
SMILES C1=CC=C2C(=C1)C=CC(=C2S(=O)(=O)O)N

Specifications

Quantity 500 g
Beilstein 613084
Merck Index 14,6406
Formula Weight 223.25
Percent Purity 98%
Chemical Name or Material 2-Aminonaphthalene-1-sulfonic acid
compliance-icons
Product Identifier
  • 2-Aminonaphthalene-1-sulfonic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2-Amino-1-naphthalenesulfonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.