missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoterephthalic acid, 99%

Product Code. 11485887
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485887 5 g 5g
11495887 25 g 25g
11405897 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485887 Supplier Thermo Scientific Alfa Aesar Supplier No. B24441.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10312-55-7 | C8H5NO4 | 179.13 g/mol

2-Aminoterephthalic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminoterephthalic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10312-55-7
Molecular Formula C8H5NO4
Molecular Weight (g/mol) 179.13
MDL Number MFCD00134536
InChI Key GPNNOCMCNFXRAO-UHFFFAOYSA-L
Synonym aminoterephthalic acid, 2-aminobenzene-1,4-dicarboxylic acid, 2-amino-1,4-benzenedicarboxylic acid, 4-carboxyanthranilic acid, 1,4-benzenedicarboxylic acid, 2-amino, 2-aminoterephthalic, aminoterephthalicacid, 2,5-dicarboxyaniline, 2-aminoterephtalic acid, timtec-bb sbb006751
PubChem CID 2724822
IUPAC Name 2-aminoterephthalic acid
SMILES NC1=CC(=CC=C1C([O-])=O)C([O-])=O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 181.15
Percent Purity 99%
Chemical Name or Material 2-Aminoterephthalic acid
compliance-icons
Product Identifier
  • 2-Aminoterephthalic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2-Aminoterephthalicacid

RUO – Research Use Only

Product Title
Select an issue

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.