missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoterephthalic acid, 98.5%, Thermo Scientific Chemicals

Product Code. p-3756839 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10399340

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10312-55-7 | C8H5NO4 | 179.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10312-55-7
Molecular Formula C8H5NO4
Molecular Weight (g/mol) 179.13
MDL Number MFCD00134536
InChI Key GPNNOCMCNFXRAO-UHFFFAOYSA-L
Synonym aminoterephthalic acid, 2-aminobenzene-1,4-dicarboxylic acid, 2-amino-1,4-benzenedicarboxylic acid, 4-carboxyanthranilic acid, 1,4-benzenedicarboxylic acid, 2-amino, 2-aminoterephthalic, aminoterephthalicacid, 2,5-dicarboxyaniline, 2-aminoterephtalic acid, timtec-bb sbb006751
PubChem CID 2724822
IUPAC Name 2-aminoterephthalic acid
SMILES NC1=CC(=CC=C1C([O-])=O)C([O-])=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 324.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 14, 558
Solubility Information Solubility in water: slightly soluble. Other solubilities: insoluble in solvents
Formula Weight 181.15
Percent Purity 99%
Physical Form Granular Powder
Chemical Name or Material 2-Aminoterephthalic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminoterephthalic acid
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.