missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiophenol, 98%

Product Code. 11493188
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493188 100 g 100g
11329679 25 g 25g
11403198 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493188 Supplier Thermo Scientific Alfa Aesar Supplier No. A18952.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 137-07-5 | C6H7NS | 125.19 g/mol

2-Aminothiophenol was used in the synthesis of 1,5-benzothiazepines derivatives by reacting with 1,3-diphenylpropenone derivatives and using aluminosilicate solid catalysts. It was also used in the synthesis of benzothiazole2 and 3-(benzothiazol-2-yl)coumarin derivatives. The ternary complexes of copper (II) with salicylidene-2-aminothiophenol (L) and glycine, alanine, valine and histidene amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminothiophenol was used in the synthesis of 1,5-benzothiazepines derivatives by reacting with 1,3-diphenylpropenone derivatives and using aluminosilicate solid catalysts. It was also used in the synthesis of benzothiazole2 and 3-(benzothiazol-2-yl)coumarin derivatives. The ternary complexes of copper (II) with salicylidene-2-aminothiophenol (L) and glycine, alanine, valine and histidene amino acids.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-07-5
Molecular Formula C6H7NS
Molecular Weight (g/mol) 125.19
MDL Number MFCD00007702
InChI Key VRVRGVPWCUEOGV-UHFFFAOYSA-N
Synonym 2-aminothiophenol, o-aminothiophenol, 2-mercaptoaniline, o-mercaptoaniline, o-aminobenzenethiol, 2-mercaptaniline, benzenethiol, 2-amino, 2-amino thiophenol, 1-amino-2-mercaptobenzene, 2-aminobenzene-1-thiol
PubChem CID 8713
IUPAC Name 2-aminobenzenethiol
SMILES NC1=CC=CC=C1S

Specifications

Melting Point 18°C to 23°C
Density 1.17
Boiling Point 234°C
Flash Point 79°C (174°F)
Odor Stench
Refractive Index 1.642
Quantity 100 g
UN Number UN3267
Beilstein 606076
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 125.19
Percent Purity 98%
Chemical Name or Material 2-Aminothiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.