missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylbenzoic Acid, 98+%

Product Code. 10112230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10112230 25 g Plastic bottle 25g
10070720 500 g Plastic bottle 500g
10677242 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10112230 Supplier Thermo Scientific Acros Supplier No. 105710250
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 85-52-9 | C14H10O3 | 226.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-52-9
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD00002472
InChI Key FGTYTUFKXYPTML-UHFFFAOYSA-N
Synonym o-benzoylbenzoic acid, benzoic acid, 2-benzoyl, 2-carboxybenzophenone, benzophenone-2-carboxylic acid, benzoic acid, o-benzoyl, o-carboxybenzophenone, benzophenone-2-carbonic acid, 2-benzoquinonecarboxylic acid, benzoylbenzoic acid, 2-phenylcarbonyl benzoic acid
PubChem CID 6813
IUPAC Name 2-benzoylbenzoic acid
SMILES OC(=O)C1=CC=CC=C1C(=O)C1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 126.0°C to 130.0°C
Color Beige to White
Boiling Point 257.0°C to 265.0°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula C6H5COC6H4CO2H
Quantity 25 g
Beilstein 10, 747
Formula Weight 226.23
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 2-Benzoylbenzoic acid, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Benzoylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.