missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyl-4-chlorophenol, 96%, Thermo Scientific Chemicals

Product Code. 15403207
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
15403207 25 g 25g
11430631 100 g 100g
11309679 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 15403207

Brand: Thermo Scientific Alfa Aesar B20112.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used in soaps, anionic detergents, cosmetics and aerosol spray products. It is a useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzyl-4-chlorophenol is used in formulating disinfectant and sanitizer products. It is also used in soaps, anionic detergents, cosmetics and aerosol spray products. It is a useful biochemical for proteomics research.

Solubility
Soluble in ethanol, alkaline solutions. Soluble in water(<1mg/mL).

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-32-1
Molecular Formula C13H11ClO
Molecular Weight (g/mol) 218.68
MDL Number MFCD00020140
InChI Key NCKMMSIFQUPKCK-UHFFFAOYSA-N
Synonym clorophene, chlorophene, clorofene, o-benzyl-p-chlorophenol, neosabenyl, septiphene, benzylchlorophenol, bio-clave, 4-chloro-2-benzylphenol, santophen 1
PubChem CID 8425
IUPAC Name 2-benzyl-4-chlorophenol
SMILES OC1=CC=C(Cl)C=C1CC1=CC=CC=C1

Specifications

Melting Point 45°C to 51°C
Density 1.19
Boiling Point 160°C to 162°C (5 mmHg)
Flash Point 185°C (365°F)
Quantity 25 g
UN Number UN3077
Beilstein 1959194
Merck Index 14,2407
Solubility Information Soluble in ethanol,alkaline solutions. Soluble in water(<1mg/ml).
Formula Weight 218.69
Percent Purity 96%
Chemical Name or Material 2-Benzyl-4-chlorophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.