missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxyethanol, 98%

Product Code. 10701365 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10701365

Brand: Thermo Scientific Acros 314650250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-08-2 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-08-2
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002868
InChI Key CUZKCNWZBXLAJX-UHFFFAOYSA-N
Synonym 2-benzyloxy ethanol, 2-benzyloxyethanol, ethanol, 2-phenylmethoxy, ethylene glycol monobenzyl ether, 2-benzyloxy-1-ethanol, benzyl cellosolve, glycol benzyl ether, ethanol, 2-benzyloxy, glycol monobenzyl ether
PubChem CID 12141
IUPAC Name 2-phenylmethoxyethanol
SMILES OCCOCC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.0700g/mL
Boiling Point 265°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2OCH2CH2OH
Refractive Index 1.5200 to 1.522
Quantity 25 g
Specific Gravity 1.07
Formula Weight 152.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Benzyloxyethanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.