missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxyphenol, 98%

Product Code. 11459326
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459326 25 g 25g
11449326 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459326 Supplier Thermo Scientific Alfa Aesar Supplier No. A19020.14
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6272-38-4 | C13H12O2 | 200.237 g/mol

2-Benzyloxyphenol is used in the synthesis of 2-(benzyloxy)hydroquinone and also used to prepare sequential polypeptides.It is used as reagent for synthesis of multidentate chelating ligands. It is also used in chemicals, pharmaceutical research and also acts a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzyloxyphenol is used in the synthesis of 2-(benzyloxy)hydroquinone and also used to prepare sequential polypeptides.It is used as reagent for synthesis of multidentate chelating ligands. It is also used in chemicals, pharmaceutical research and also acts a pharmaceutical intermediate.

Solubility
Soluble in alcohol, benzene, and diethyl ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6272-38-4
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00002186
InChI Key CCZCXFHJMKINPE-UHFFFAOYSA-N
Synonym 2-benzyloxy phenol, 2-benzyloxyphenol, phenol, 2-phenylmethoxy, benzyloxyphenol, benzyl o-hydroxyphenyl ether, 2-phenylmethoxy phenol, 2-benzyloxy-phenol, o-benzyloxy phenol
PubChem CID 80459
IUPAC Name 2-phenylmethoxyphenol
SMILES C1=CC=C(C=C1)COC2=CC=CC=C2O

Specifications

Density 1.143
Flash Point >110°C (230°F)
Refractive Index 1.591
Quantity 25 g
Solubility Information Soluble in alcohol,benzene,and diethyl ether. Insoluble in water.
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material 2-Benzyloxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.