missing translation for 'onlineSavingsMsg'
Learn More

2-(Bis(3,5-dimethylphenyl)phosphino)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10567020 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 10567020

Brand: Thermo Scientific Acros 435142500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 669091-00-3
CAS Min % 96.0
CAS Max % 100.0
Melting Point 109.5°C to 111.0°C
Color Yellow
Elemental Analysis 76.56 to 82.94% C
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C23H23OP
MDL Number MFCD15143662
Quantity 250 mg
Synonym 2-bis 3,5-dimethylphenyl phosphino benzaldehyde, 2-bis 3,5-dimethylphenyl phosphanyl benzaldehyde, acmc-20ao6m, 2-bis 3,5-dimethylphenyl phosphino benzaldehyde,min
InChI Key PQIAXRGPKDEYLW-UHFFFAOYSA-N
SMILES CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=CC=CC=C1C=O
IUPAC Name 2-bis(3,5-dimethylphenyl)phosphanylbenzaldehyde
Molecular Weight (g/mol) 346.41
PubChem CID 71310677
Formula Weight 346.41
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-(Bis(3, 5-Dimethylphenyl)phosphino)benzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Bis(3,5-dimethylphenyl)phosphino)benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.