missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)benzeneboronic acid, 96%, Thermo Scientific™

Product Code. 15499187
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499187 250 mg 250mg
15489187 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499187 Supplier Thermo Scientific Alfa Aesar Supplier No. H52448.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115377-94-1
Molecular Formula C11H16BNO4
Molecular Weight (g/mol) 237.062
MDL Number MFCD02179450
InChI Key RMEIKYSECODAHJ-UHFFFAOYSA-N
Synonym 2-boc-aminophenyl boronic acid, 2-tert-butoxycarbonyl amino phenyl boronic acid, n-tert-butoxycarbonyl-2-amino-1-phenylboronic acid, 2-n-boc-amino phenylboronic acid, 2-tert-butoxycarbonylamino phenylboronic acid, n-tert-butoxycarbonyl 2-amino-1-phenylboronic acid, 2-2-methylpropan-2-yl oxycarbonylamino phenyl boronic acid, 2-tert-butoxycarbonyl amino phenylboronic acid, 2-t-butoxycarbonylamino phenylboronic acid, carbamic acid, n-2-boronophenyl-, 1,1-dimethylethyl ester
PubChem CID 2773226
IUPAC Name [2-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid
SMILES B(C1=CC=CC=C1NC(=O)OC(C)(C)C)(O)O

Specifications

Melting Point 123°C to 125°C
Quantity 250 mg
Formula Weight 237.06
Percent Purity 96%
Chemical Name or Material 2-(Boc-amino)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.