missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)benzeneboronic acid pinacol ester, 97%

Product Code. 11432210
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432210 1 g 1g
11442210 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432210 Supplier Thermo Scientific Alfa Aesar Supplier No. H29020.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 159624-15-4 | C17H26BNO4 | 319.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159624-15-4
Molecular Formula C17H26BNO4
Molecular Weight (g/mol) 319.208
MDL Number MFCD03411943
InChI Key LVHGGVGVAUJQBB-UHFFFAOYSA-N
Synonym tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-amino benzeneboronic acid pinacol ester, 2-n-boc-amino phenylboronic acid pinacol ester, tert-butyl-n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenylboronic acid pinacol ester, tert-butyl n-2-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenyl boronic acid, pinacol ester, 2-boc-amino phenylboronic acid pinacol ester, 2-tert-butoxycarbonylamino phenylboronic acid, pinacol ester
PubChem CID 2773231
IUPAC Name tert-butyl N-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2NC(=O)OC(C)(C)C

Specifications

Melting Point 98°C to 100°C
Quantity 1 g
Formula Weight 319.21
Percent Purity 97%
Chemical Name or Material 2-(Boc-amino)benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.