missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)pyridine, 95%

Product Code. 15483977
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483977 1 g 1g
15403987 250 mg 250mg
15493977 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15483977 Supplier Thermo Scientific Alfa Aesar Supplier No. H30741.03
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 38427-94-0 | C10H14N2O2 | 194.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38427-94-0
Molecular Formula C10H14N2O2
Molecular Weight (g/mol) 194.234
MDL Number MFCD03411622
InChI Key ORUGTGTZBRUQIT-UHFFFAOYSA-N
Synonym 2-boc-amino pyridine, tert-butyl pyridin-2-ylcarbamate, 2-tert-butoxycarbonylamino pyridine, tert-butyl n-pyridin-2-yl carbamate, tert-butyl n-2-pyridyl carbamate, pyridin-2-yl-carbamic acid tert-butyl ester, carbamic acid, 2-pyridinyl-, 1,1-dimethylethyl ester, carbamic acid, n-2-pyridinyl-, 1,1-dimethylethyl ester, pubchem17730, boc-2-aminopyridine
PubChem CID 11206349
IUPAC Name tert-butyl N-pyridin-2-ylcarbamate
SMILES CC(C)(C)OC(=O)NC1=CC=CC=N1

Specifications

Melting Point 92°C to 94°C
Quantity 1 g
Formula Weight 194.23
Percent Purity 95%
Chemical Name or Material 2-(Boc-amino)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.