missing translation for 'onlineSavingsMsg'
Learn More

2-bromo-1-(2-phenoxyphenyl)ethanone, Thermo Scientific™

Product Code. 10307644
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10307644 250 mg Amber Glass Bottle 250mg
10164244 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10307644 Supplier Thermo Scientific Maybridge Supplier No. CC49905CB
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 94402-42-3
Molecular Formula C14H11BrO2
Molecular Weight (g/mol) 291.144
MDL Number MFCD08435906
InChI Key UWRDQDVDGPRCKR-UHFFFAOYSA-N
Synonym 2-bromo-1-2-phenoxyphenyl ethanone, 2-bromo-1-2-phenoxyphenyl ethan-1-one, 2-phenoxy phenacyl bromide, 2-bromoacetyl diphenyl ether
PubChem CID 13074046
IUPAC Name 2-bromo-1-(2-phenoxyphenyl)ethanone
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2C(=O)CBr

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 291.14
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(2-phenoxyphenyl)ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.