missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3,5-trimethyl-4-nitrobenzene, 90+%

Product Code. 11391827
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11391827 1 g 1g
11301837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391827 Supplier Thermo Scientific Alfa Aesar Supplier No. L05107.03
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 90561-85-6 | C9H10BrNO2 | 244.088 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90561-85-6
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.088
MDL Number MFCD00015924
InChI Key IUFDRRDWXUSFBG-UHFFFAOYSA-N
Synonym 3-bromo-2,4,6-trimethylnitrobenzene, 2-bromo-4-nitro-1,3,5-trimethylbenzene, 1-bromo-3-nitro-2,4,6-trimethylbenzene, 4-bromo-2-nitromesitylene, 4-bromo-1,3,5-trimethyl-2-nitrobenzene, 2-bromanyl-1,3,5-trimethyl-4-nitro-benzene
PubChem CID 2757200
IUPAC Name 2-bromo-1,3,5-trimethyl-4-nitrobenzene
SMILES CC1=CC(=C(C(=C1[N+](=O)[O-])C)Br)C

Specifications

Melting Point 56°C to 62°C
Boiling Point 106°C to 107°C (1.3 mmHg)
Quantity 1 g
Beilstein 1958703
Sensitivity Light sensitive
Formula Weight 244.09
Percent Purity ≥90%
Chemical Name or Material 2-Bromo-1,3,5-trimethyl-4-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.